Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(I) Chloride - Bis(Lithium Chloride) Complex

Catalog No. 70174250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-174-250ML 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 70-174-250ML Supplier Sigma Aldrich Supplier No. 70174250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Solution used in Cu(I) transmetallation for amination reaction

Specifications

CAS 121898-64-4
Melting Point 174°C to 179°C
Boiling Point 165°C (400 mmHg, lit.), 190°C to 191°C
Molecular Formula C27H33O3P
Linear Formula C27H33O3P
Quantity 500 mg
MDL Number MFCD03840342
Molecular Weight (g/mol) 436.52 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.