Learn More
Strem, An Ascensus Company CAS# 544-92-3. 100g. Copper(I) cyanide, 99%. MFCD00010975
SDP
Catalog No. 5090114875
Encompass
CAS# 544-92-3. 100g. Copper(I) cyanide, 99%. MFCD00010975. Molecular Weight: 89.56. Molecular Formula: CuCN. Color/form: off-white to pale green pwdr. Strem# 93-2916. http://www.strem.com/catalog/v/93-2916/
Catalog No. 50-901-14875 Supplier Strem, An Ascensus Company Supplier No. 932916100G
CAS# 544-92-3. 100g. Copper(I) cyanide, 99%. MFCD00010975. Molecular Weight: 89.56. Molecular Formula: CuCN. Color/form: off-white to pale green pwdr. Strem# 93-2916. http://www.strem.com/catalog/v/93-2916/