Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(I) Cyanide Di(Lithium Chloride) Complex Solution

Catalog No. 69725750ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-725-750ML 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 69-725-750ML Supplier Sigma Aldrich Supplier No. 69725750ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Copper(I) cyanide di(lithium chloride) complex can be used for the synthesis of organocopper(I) reagents by transmetalation with organozinc and Grignard reagents. It is also a useful precursor to prepare organocuprate(I) reagents.

Specifications

CAS 458532-88-2
Melting Point 171°C to 177°C
Molecular Formula C22H44P2Rh
Linear Formula C22H44P2Rh
MDL Number MFCD11973806
Quantity 100 mg
Synonym (R)-TCFP-Rh; TCFP; R-[Rh cod TCFP]BF4
Molecular Weight (g/mol) 473.44 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.