Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) cyanide

Catalog No. AA12135A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 x 2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12135A1 1 kg
AA1213530 250 g
AA12135F3 2 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA12135A1 Supplier Thermo Scientific Chemicals Supplier No. 012135A1
Add to Cart

For Electroplating Cu or Fe, Polymerization Catalyst | CAS: 544-92-3 | CCuN | 89.564 g/mol

Copper(I) cyanide is commonly used as a polymerization catalyst. The compound is useful in organic synthesis, as a catalyst, reagent in the preparation of nitriles(sandmeyer reaction). It is also used in electroplating copper and iron, silver plating, brass plating, and copper-tin alloy plating. It is used to make phthalocyanine dyes and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-92-3
Molecular Formula CCuN
Molecular Weight (g/mol) 89.564
MDL Number MFCD00010975
InChI Key DOBRDRYODQBAMW-UHFFFAOYSA-N
Synonym copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
PubChem CID 11009
IUPAC Name copper(1+);cyanide
SMILES [C-]#N.[Cu+]

Specifications

Melting Point 473°C
Color White
Physical Form Powder
Quantity 1 kg
Beilstein 3587244
Solubility Information Practically insoluble in water and alcohol. Soluble in ammonium hydroxide,aqueous ammonia,pyridine and N-methylpyrrolidone.
Formula Weight 89.56
Odor Characteristic
Density 2.92
Chemical Name or Material Copper(I) cyanide
Hazard Category H300+H310+H330
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 208-883-6
RTECSNumber GL7150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.