Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(I) iodide

Catalog No. 2155541KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-555-41KG 1 kg
21-555-4100G 100 g
21-555-425G 25 g
21-555-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-555-41KG Supplier Sigma Aldrich Supplier No. 2155541KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Copper(I) iodide

Specifications

Boiling Point 1290°C
Melting Point 605°C
CAS 7681-65-4
Quantity 1 kg
Molecular Formula CuI
Linear Formula CuI
MDL Number MFCD00010978
Synonym Cuprous iodide
Molecular Weight (g/mol) 190.45
Percent Purity 99.999% trace metals basis
Density 5.62 g/mL (at 25°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.