Learn More
Strem, An Ascensus Company CAS# 7681-65-4. 100g. Copper(I) iodide, 98%. MFCD00010978
SDP
Catalog No. 5090114890
Encompass
CAS# 7681-65-4. 100g. Copper(I) iodide, 98%. MFCD00010978. Molecular Weight: 190.45. Molecular Formula: CuI. Color/form: off-white to pale brown pwdr. Strem# 93-2936. http://www.strem.com/catalog/v/93-2936/
Catalog No. 50-901-14890 Supplier Strem, An Ascensus Company Supplier No. 932936100G
CAS# 7681-65-4. 100g. Copper(I) iodide, 98%. MFCD00010978. Molecular Weight: 190.45. Molecular Formula: CuI. Color/form: off-white to pale brown pwdr. Strem# 93-2936. http://www.strem.com/catalog/v/93-2936/