Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) iodide, Puratronic™, 99.998% (metals basis)

Catalog No. AA4315306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4315306 5 g
AA4315314 25 g
AA4315322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4315306 Supplier Thermo Scientific Chemicals Supplier No. 04315306
Add to Cart

missing translation for 'validMainframeMsg'

Light, Air and Moisture Sensitive | CAS: 7681-65-4 | CuI

Copper(I) iodide is used as a catalyst in organic synthesis, cathode ray tube coating, cloud seeding, and as temperature indicator with HgI2. It catalyzes the conversion of aryl, heteroaryl, and vinyl bromides into the corresponding iodides, which are employed in coupling reactions such as Suzuki, Stille, Sonogashira, Heck and Ullmann reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-65-4
Molecular Formula CuI
MDL Number MFCD00010978
Synonym cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g

Specifications

Boiling Point ∼1290°C
Melting Point 605°C
Color Brown to White
Physical Form Chunks and/or Powder
Quantity 5 g
Odor Weak odor
Refractive Index 2.346
Sensitivity Air, light, and moisture sensitive
Appearance White to pale brown chunks/powder
Assay Percent Range 99.998% (Metal basis)
Density 5.63 g/cm3
Solubility Information Copper(I) iodide is soluble in ammonia,thiosulfate,sodium and potassium iodide solutions. Insoluble in water and dilute acids.
Formula Weight 190.45
Chemical Name or Material Copper(I) Iodide
Show More Show Less
Hazard Category H302-H315-H317-H318-H372
Precautionary Statement P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P314-P330-P333+P313-P362-P501c
DOTInformation DOT Class: 9, Packing Group: III
EINECSNumber 231-674-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.