Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) Oxide, 97%, Solstice

Catalog No. 60045171 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-171 2 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-171 Supplier Solstice Supplier No. 2088252KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Powder, <5 μm, 97%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-39-1
MDL Number MFCD00010974

Specifications

Melting Point 1230°C
Color Brown or Red
pH 8.0 (at 100g/L 20°C)
Physical Form Powder
Quantity 2 kg
Linear Formula Cu2O
UN Number UN3077
Molecular Weight (g/mol) 143.09
Formula Weight 143.09g/mol
Percent Purity 97%
Packaging Poly Bottle
Density 6g/mL at 25°C
Chemical Name or Material Copper(I) Oxide
Show More Show Less
Hazard Statement P261-P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302 + H332-H319-H410
DOTInformation 9/PGIII
EINECSNumber 215-270-7
RTECSNumber GL8050000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.