Learn More
Strem, An Ascensus Company CAS# 1317-39-1. 25g. Copper(I) oxide (99.9%-Cu). MFCD00010974
SDP
Catalog No. 5090114934
Encompass
CAS# 1317-39-1. 25g. Copper(I) oxide (99.9%-Cu). MFCD00010974. Molecular Weight: 143.08. Molecular Formula: Cu2O. Color/form: red-brown pwdr. Strem# 93-2981. http://www.strem.com/catalog/v/93-2981/
Catalog No. 50-901-14934 Supplier Strem, An Ascensus Company Supplier No. 93298125G
CAS# 1317-39-1. 25g. Copper(I) oxide (99.9%-Cu). MFCD00010974. Molecular Weight: 143.08. Molecular Formula: Cu2O. Color/form: red-brown pwdr. Strem# 93-2981. http://www.strem.com/catalog/v/93-2981/