Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) oxide, 99% (metals basis)

Catalog No. AA4018836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4018836 500 g
AA4018814 25 g
AA4018822 100 g
AA40188A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4018836 Supplier Thermo Scientific Chemicals Supplier No. 04018836
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1317-39-1 | Cu2O

Used in antifouling, marine paint, brazing paste, antifungal treatment for plant seeds. Used as a hydrogen scavenger in copper brass and bronze and high grade lubricants. Finds uses in glass and ceramic pigments, ferrite magnets, catalyst in organic synthesis, dyes and dye intermediates and as catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-39-1
Molecular Formula Cu2O
MDL Number MFCD00010974

Specifications

Boiling Point 1800°C (decomposition)
Melting Point 1235°C
Color Purple-Red
Physical Form Powder
Quantity 500 g
Odor Odorless
Refractive Index 2.705
Sensitivity Air and moisture sensitive
Appearance Purple-red powder
Assay Percent Range 99% (Metal basis)
Density 6 g/cm3
Solubility Information It is insoluble in water and organic solvents.
Formula Weight 143.08
Chemical Name or Material Copper(I) oxide
Show More Show Less
Hazard Category H302+H332-H318
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c
DOTInformation DOT Class: 9, Packing Group: III
EINECSNumber 215-270-7
RTECSNumber GL8050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.