Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) Oxide, Solstice

Catalog No. 6000435 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 kg
6 x 2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-004-35 2.5 kg
60-044-764 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-004-35 Supplier Solstice Supplier No. 128412.5KG
Add to Cart
Add to Cart

≥86% Cu basis, red

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-39-1
Molecular Formula Cu2O
MDL Number MFCD00010974

Specifications

Color Red
Quantity 2.5 kg
UN Number 3077
Formula Weight 143.09g/mol
Percent Purity ≥86% (Cu basis)
Packaging Plastic Bottle
Chemical Name or Material Copper(I) Oxide
Hazard Statement P261-P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302 + H332-H319-H410
EINECSNumber 215-270-7
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.