Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(I) Thiophene-2-Carboxylate

Catalog No. 6825001G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-250-01G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-250-01G Supplier Sigma Aldrich Supplier No. 6825001G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant or reagent involved in:Studies of xenobiotic response to herbicide safener derivativesSynthesis of functionalized BODIPY dye analogsOrthogonal cross-coupling reactionsPreparation of parent borondipyrromethene systemCopper mediated cross-coupling

Specifications

CAS 651042-82-9
Molecular Formula C15H32O7S
Linear Formula C15H32O7S
MDL Number MFCD08274612
Quantity 250 mg
Synonym mPEG thiol
Molecular Weight (g/mol) 356.48 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.