Learn More
Strem, An Ascensus Company CAS# 149-11-1. 25g. Copper(II) 2-ethylhexanoate (solvent free - 16-19% Cu). MFCD00015695
SDP
Catalog No. 5090111311
Encompass
CAS# 149-11-1. 25g. Copper(II) 2-ethylhexanoate (solvent free - 16-19% Cu). MFCD00015695. Molecular Weight: 349.96. Molecular Formula: Cu[OOCCH(C2H5)C4H9]2. Color/form: blue pwdr. Strem# 29-2941. http://www.strem.com/catalog/v/29-2941/
Catalog No. 50-901-11311 Supplier Strem, An Ascensus Company Supplier No. 29294125G
CAS# 149-11-1. 25g. Copper(II) 2-ethylhexanoate (solvent free - 16-19% Cu). MFCD00015695. Molecular Weight: 349.96. Molecular Formula: Cu[OOCCH(C2H5)C4H9]2. Color/form: blue pwdr. Strem# 29-2941. http://www.strem.com/catalog/v/29-2941/