Learn More
Medchemexpress LLC Copper(II) acetate | 142-71-2 | MFCD00008690 | 100.0% | 181.63 | C4H6CuO4 | 1g
SDP
Catalog No. 5000267679
Encompass_Preferred
Copper(II) acetate 99 99% trace metals basis is a dinuclear metal complex that can be used a source of copper and oxidizing agent in inorganic synthesis and as a catalyst in organic synthesis[1]
Catalog No. 50-002-67679 Supplier Medchemexpress LLC Supplier No. HYY1102A1G
Copper(II) acetate 99 99% trace metals basis is a dinuclear metal complex that can be used a source of copper and oxidizing agent in inorganic synthesis and as a catalyst in organic synthesis[1]