Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(II) acetate

Catalog No. 5174535G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-745-35G 5 g
51-745-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 51-745-35G Supplier Sigma Aldrich Supplier No. 5174535G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Copper acetate (Cu (OAc)2) occurs in dimeric structure in solid state and in non-aqueous solution. Anhydrous Cu (OAc)2 may be manufactured by heating or refluxing the monohydrate Cu (OAc)2 at 90oC and then washing the insoluble product with diethyl ether.Copper acetate forms complexes which may be used in biochemical reactions such as; DNA extraction, binding ribose hydroxyls of purine and pyrimidine nucleosides. A study has shown Cu (OAc)2 to fix and remove tannins in samples from Pyrus yielding high quality DNA. The complexes have been studied for anticancer, anti-bacterial and antifungal activities.

Specifications

CAS 142-71-2
Molecular Formula C4H6CuO4
Linear Formula Cu(CO2CH3)2
Quantity 5 g
MDL Number MFCD00008690
Synonym Cupric acetate
Molecular Weight (g/mol) 181.63
Percent Purity 99.99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.