Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem Copper(II) acetate | 95% | CAS 142-71-2 | C4H8CuO4 | 1kg

Catalog No. 5001023595
Encompass_Preferred

Copper(II) acetate | 95% | CAS 142-71-2 | C4H8CuO4 | 1kg

Catalog No. 50-010-23595 Supplier Aobchem Supplier No. A225721KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Copper(II) acetate | 95% | CAS 142-71-2 | C4H8CuO4 | 1kg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.