Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) acetate, 99%, pure, anhydrous

Catalog No. AC405370010
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405370010 1 kg Plastic bottle
AC405370500 50 g Plastic bottle
AC405372500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC405370010 Supplier Thermo Scientific Chemicals Supplier No. 405370010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 142-71-2 | C4H6CuO4 | 181.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-71-2
Molecular Formula C4H6CuO4
Molecular Weight (g/mol) 181.64
InChI Key OPQARKPSCNTWTJ-UHFFFAOYSA-L
Synonym copper ii acetate, cupric acetate, copper diacetate, copper acetate, venus copper, copper 2+ diacetate, cupric diacetate, neutral verdigris, crystals of venus
PubChem CID 8895
IUPAC Name copper;diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Cu+2]

Specifications

Melting Point 115.0°C
Color Blue to Green
Physical Form Crystalline Powder
Quantity 1 kg
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 99%
Linear Formula (CH3CO2)2Cu
Fieser 01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 15,99
Merck Index 15,2614
Solubility Information Solubility in water: soluble
Formula Weight 181.64
Percent Purity 99%
Grade Pure
Chemical Name or Material Copper(II) acetate
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 205-553-3

RUO – Research Use Only

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.