Learn More
Strem, An Ascensus Company COPPER(II) ACETATE MONOHYD-50G
SDP
Catalog No. 5090114855
Encompass
Copper (II) acetate monohydrate, 98+% (ACS); Size: 50g; [6046-93-1]; Cu(OOCCH3)2.H2O; Mol Wt:181.64 (199.65); Color & Form: green to blue pwdr.
Catalog No. 50-901-14855 Supplier Strem, An Ascensus Company Supplier No. 93290150G
Copper (II) acetate monohydrate, 98+% (ACS); Size: 50g; [6046-93-1]; Cu(OOCCH3)2.H2O; Mol Wt:181.64 (199.65); Color & Form: green to blue pwdr.