Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (II) Acetate, Monohydrate
SCI_ED

Catalog No. S25276A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25276A 500 g Lab Grade
S25276 100 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25276A

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 6046-93-1
Molecular Formula C4H8CuO5
Molecular Weight (g/mol) 199.65
MDL Number MFCD00149570
InChI Key NWFNSTOSIVLCJA-UHFFFAOYSA-L
Synonym copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
PubChem CID 165397
IUPAC Name copper(2+) diacetate hydrate
SMILES O.[Cu++].CC([O-])=O.CC([O-])=O

Specifications

Color Blue-Green
Quantity 500 g
Assay Percent Range >93%
Linear Formula (CH3CO2)2Cu·H2O
Formula Weight 199.65
Grade Laboratory
Packaging Lab Grade
Density 1.88g/mL
Chemical Name or Material Copper (II) Acetate, Monohydrate
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 9, III, : Miscellaneous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.