Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(II) acetate monohydrate

Catalog No. 22960110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-960-110G 10 g
22-960-150G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-960-110G Supplier Sigma Aldrich Supplier No. 22960110G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Copper (II) acetate monohydrate is soluble in water and slightly soluble in methanol, diethylether and acetone. It can be synthesized by reacting acetic acid with copper (II) carbonate or copper(II) hydroxide or copper (II) oxide. Large scale commercial production is undertaken by placing copper metal in the presence of air and refluxing acetic acid. As a transition metal acetate, it can be used to form oxide nanoparticles by sonochemical methods.Used in the synthesis of copper oxide nanoparticles, Cu2ZnSnS4 films, which find applications in solar cells. It may be used in Zinc Oxide varistors, as a catalyst in the polymerization of organic materials and as a mordant in the dying of textiles. Industrially, it may be used as a ceramic pigment and as a fungicide.

Specifications

CAS 6046-93-1
Molecular Formula C4H6CuO4 -+ H2O
Linear Formula Cu(CO2CH3)2 -+ H2O
MDL Number MFCD00149570
Quantity 10 g
Synonym Cupric acetate monohydrate
Molecular Weight (g/mol) 199.65
Percent Purity 99.99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.