Learn More
Copper(II) acetate monohydrate, 98+%
Description
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.
Solubility
Soluble in water and alcohol. Slightly soluble in ether and glycerine.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 115°C |
| Quantity | 250 g |
| Density | 1.882 g/mL |
| Linear Formula | Cu(OOCCH3)2·H2O |
| UN Number | UN3077 |
| Merck Index | 14,2624 |
| Solubility Information | Soluble in water and alcohol. Slightly soluble in ether and glycerine. |
| Formula Weight | 199.65 (181.64 Anhydrous) |
| Percent Purity | ≥98% |
| Chemical Name or Material | Copper(II) acetate monohydrate |
Safety and Handling
| Hazard Category | H302-H314-H335-H373 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 205-553-3 |
| RTECSNumber | AG3500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.