Learn More
Medchemexpress LLC Copper(II) acetylacetonate | 13395-16-9 | MFCD00000016 | ≥99.0% | 261.76 g/mol | C10H14CuO4 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW0348011KG
Encompass_Preferred
Copper(II) acetylacetonate is a homoleptic acetylacetonate complex of copper(II) used as a catalyst and precursor in organic synthesis, materials preparation, and coordination chemistry. It is a purple-blue solid with a reported molecular weight of 261.76 g/mol and high purity suitable for research applications.
  • High purity suitable for research and synthesis.
  • Purple-blue solid that is stable and easy to handle under normal laboratory conditions.
  • Commonly used as a catalyst in oxidation, cross-coupling, and polymerization reactions.
  • Well-characterized composition (C10H14CuO4) and molecular weight.
  • Available in multiple pack sizes for laboratory and scale-up use.
Catalog No. 50-002-99437