Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Copper(II) acetylacetonate | 13395-16-9 | MFCD00000016 | >99.0% | 261.76 g·mol⁻¹ | C10H14CuO4 | 250 G
SDP

Catalog No. 5000303724
Encompass_Preferred

Copper(II) acetylacetonate is the homoleptic acetylacetonate complex of copper(II) commonly used as a catalyst and as a precursor in materials synthesis. Supplied as a high-purity solid, it is suitable for use in organic synthesis, polymerization, and preparation of metal-organic frameworks and nanomaterials.

  • Used as a catalyst in oxidation, cross-coupling, and polymerization reactions.
  • Precursor for metal-organic frameworks and nanomaterials synthesis.
  • High purity (≥99.0%) for reproducible results.
  • Solid powder form, commonly bluish or greenish in appearance.
  • Molecular formula C10H14CuO4; molecular weight 261.76 g·mol⁻¹.
  • CAS number 13395-16-9 for unambiguous identification.

Catalog No. 50-003-03724 Supplier Medchemexpress LLC Supplier No. 250GHYW034801
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Copper(II) acetylacetonate is the homoleptic acetylacetonate complex of copper(II) commonly used as a catalyst and as a precursor in materials synthesis. Supplied as a high-purity solid, it is suitable for use in organic synthesis, polymerization, and preparation of metal-organic frameworks and nanomaterials.

  • Used as a catalyst in oxidation, cross-coupling, and polymerization reactions.
  • Precursor for metal-organic frameworks and nanomaterials synthesis.
  • High purity (≥99.0%) for reproducible results.
  • Solid powder form, commonly bluish or greenish in appearance.
  • Molecular formula C10H14CuO4; molecular weight 261.76 g·mol⁻¹.
  • CAS number 13395-16-9 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.