Learn More
Medchemexpress LLC Copper(II) acetylacetonate | 13395-16-9 | MFCD00000016 | ≥99.0% | 261.76 g·mol⁻¹ | C10H14CuO4 | 500 G
SDP
Catalog No. 5000299301
Encompass_Preferred
Copper(II) acetylacetonate is the homoleptic acetylacetonate complex of copper(II), used as a copper source and catalyst in organic synthesis, materials research, and coordination chemistry. It is supplied as a stable solid suitable for laboratory handling and formulation into reaction mixtures.
  • High-purity solid copper complex for research and synthesis.
  • Effective catalyst for oxidation, cross-coupling, and polymerization reactions.
  • Soluble in common organic solvents for homogeneous reaction conditions.
  • Provides a convenient copper source for materials and coordination chemistry.
  • Stable when stored protected from light and moisture under recommended conditions.
Catalog No. 50-002-99301 Supplier Medchemexpress LLC Supplier No. HYW034801500G
Copper(II) acetylacetonate is the homoleptic acetylacetonate complex of copper(II), used as a copper source and catalyst in organic synthesis, materials research, and coordination chemistry. It is supplied as a stable solid suitable for laboratory handling and formulation into reaction mixtures.
  • High-purity solid copper complex for research and synthesis.
  • Effective catalyst for oxidation, cross-coupling, and polymerization reactions.
  • Soluble in common organic solvents for homogeneous reaction conditions.
  • Provides a convenient copper source for materials and coordination chemistry.
  • Stable when stored protected from light and moisture under recommended conditions.