Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (II) Bromide
SCI_ED

Catalog No. S25277A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25277A 100 g Reagent Grade
S25277 25 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25277A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7789-45-9
Synonym Cupric Bromide

Specifications

Color Blue-Gray
Quantity 100 g
Assay Percent Range >99%
Linear Formula CuBr2
Formula Weight 223.37
Grade Reagent
Packaging Reagent Grade
Density 4.77g/mL at 20°C
Chemical Name or Material Copper (II) Bromide
Safety handling Check Alert storage code Red
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.