Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strem, An Ascensus Company Copper(II) Bromide, 99%, 50g
SDP

Catalog No. 5090114863
Encompass

Copper(II) Bromide, 99%, 7789-45-9, MFCD00010970, Br2Cu, 223.37, 50g

Catalog No. 50-901-14863 Supplier Strem, An Ascensus Company Supplier No. 93290850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Copper(II) Bromide, 99%, 7789-45-9, MFCD00010970, Br2Cu, 223.37, 50g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.