Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (II) Carbonate
SCI_ED

Catalog No. S25278 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Powder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25278 500 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25278
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 12069-69-1
Synonym Cupric Carbonate Basic, Copper; Copper (II) Hydroxide Carbonate

Specifications

Quantity 500 g
Linear Formula CuCO3·Cu(OH)2
Formula Weight 221.11
Packaging Powder
Density 4g/mL
Chemical Name or Material Copper (II) Carbonate
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.