Learn More
Strem, An Ascensus Company CAS# 12069-69-1. 500g. Copper(II) carbonate, basic. MFCD00010976
SDP
Catalog No. 5090114867
Encompass
CAS# 12069-69-1. 500g. Copper(II) carbonate, basic. MFCD00010976. Molecular Weight: 221.11. Molecular Formula: CuCO3∙Cu(OH)2. Color/form: green to blue pwdr. Strem# 93-2911. http://www.strem.com/catalog/v/93-2911/
Catalog No. 50-901-14867 Supplier Strem, An Ascensus Company Supplier No. 932911500G
CAS# 12069-69-1. 500g. Copper(II) carbonate, basic. MFCD00010976. Molecular Weight: 221.11. Molecular Formula: CuCO3∙Cu(OH)2. Color/form: green to blue pwdr. Strem# 93-2911. http://www.strem.com/catalog/v/93-2911/