Learn More
Strem, An Ascensus Company CAS# 7447-39-4. 100g. Copper(II) chloride, anhydrous, min. 98%. MFCD00010972
SDP
Catalog No. 5090114869
Encompass
CAS# 7447-39-4. 100g. Copper(II) chloride, anhydrous, min. 98%. MFCD00010972. Molecular Weight: 134.44. Molecular Formula: CuCl2. Color/form: reddish-brown pwdr. Strem# 93-2912. http://www.strem.com/catalog/v/93-2912/
Catalog No. 50-901-14869 Supplier Strem, An Ascensus Company Supplier No. 932912100G
CAS# 7447-39-4. 100g. Copper(II) chloride, anhydrous, min. 98%. MFCD00010972. Molecular Weight: 134.44. Molecular Formula: CuCl2. Color/form: reddish-brown pwdr. Strem# 93-2912. http://www.strem.com/catalog/v/93-2912/