Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (II) Chloride Solution, 1M
SCI_ED

Chemical Identifiers

CAS 10125-13-0 , 7732-18-5
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
PubChem CID 61482
ChEBI CHEBI:86318
IUPAC Name copper(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Cu++]
View More Specs

Products 2
Catalog Number Quantity Packaging Price Quantity & Availability  
Catalog Number Quantity Packaging Price Quantity & Availability  
S25280
SCI_ED
SDS View Documents
50 mL 1M N/A N/A N/A
S25280A
SCI_ED
SDS View Documents
500 mL 1M N/A N/A N/A
Description

Description

This product is for educational use - no C of A is available.

Specifications

Chemical Identifiers

10125-13-0 , 7732-18-5
170.48
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
61482
copper(2+) dihydrate dichloride
Cl2CuH4O2
MFCD00149674
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
CHEBI:86318
O.O.[Cl-].[Cl-].[Cu++]

Specifications

Blue-Green
<2%w/v
MFCD00149674
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
copper(2+) dihydrate dichloride
61482
Liquid
10125-13-0 , 7732-18-5
Cl2CuH4O2
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
O.O.[Cl-].[Cl-].[Cu++]
170.48
CHEBI:86318
Copper (II) Chloride Solution, 1M, 1.0M
Videos
Safety and Handling

Safety and Handling

ShelfLife : 1 year

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.