Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (II) Chloride Solution, 1M
SCI_ED

Catalog No. S25280A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
Packaging:
1M
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25280A 500 mL 1M
S25280 50 mL 1M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25280A

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10125-13-0 , 7732-18-5
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
PubChem CID 61482
ChEBI CHEBI:86318
IUPAC Name copper(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Color Blue-Green
Assay Percent Range <2%w/v
Packaging 1M
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Copper (II) Chloride Solution, 1M, 1.0M
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.