Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper (II) Chloride Solution
SCI_ED

Catalog No. S99603 Shop All Aldon Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S99603 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S99603 Supplier Aldon Corporation Supplier No. CC0507CONC

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7447-39-4 , 7732-18-5
Synonym Cupric chloride

Specifications

Quantity 50 mL
Chemical Name or Material Copper(II) Chloride Solution, 1.0M
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.