Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) chloride, ultra dry, 99.995% (metals basis)

Catalog No. AA3567306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3567306 5 g
AA3567314 25 g
AA3567322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3567306 Supplier Thermo Scientific Chemicals Supplier No. 03567306
Add to Cart

Air Sensitive and Hygroscopic. >300°C partial decomposition to CuCl and Cl2 | CAS: 7447-39-4 | CuCl2

Copper(II) chloride is used as a co-catalyst with palladium(II) chloride used in the Wacker process to prepare ethanol from ethane. It acts as an effective catalyst for the tetrahydropyranylation of alcohols. It is involved in the production of vinyl chloride and dichloroethane as well as in the chlorination of aromatic hydrocarbons. It acts as a deodorizing and desulfurizing agent in petroleum industry, mordant in textiles industry and in electrotyping baths for plating copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-39-4
Molecular Formula CuCl2
MDL Number MFCD00010972

Specifications

Boiling Point 993°C (decomposition)
Melting Point 620°C
Color Brown-Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.995% (Metal basis)
Solubility Information Soluble in water,alcohol,acetone and hot sulfuric acid.
Formula Weight 134.45
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Yellow-brown powder
Density 3.386 g/cm3
Chemical Name or Material Copper(II) chloride
Show More Show Less
Hazard Category H302+H312-H315-H318
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 8, Packing Group: III
EINECSNumber 231-210-2
RTECSNumber GL7000000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.