Learn More
Medchemexpress LLC Copper II diethyldi 1g
SDP
Catalog No. 5000206124
Encompass_Preferred
Copper(II) diethyldithiocarbamate has anticancer activity Copper(II) diethyldithiocarbamate can be synthesized from monovalent copper-cysteamine and disulfiram[1]
Catalog No. 50-002-06124 Supplier Medchemexpress LLC Supplier No. HYW1115811G
Copper(II) diethyldithiocarbamate has anticancer activity Copper(II) diethyldithiocarbamate can be synthesized from monovalent copper-cysteamine and disulfiram[1]