Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper(II) ethoxide, 98%

Manufacturer:  Alfa Aesar™ 4631614

Quantity 25g

 View more versions of this product

Catalog No. AA4631614


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2850-65-9
Cu(OC2H5)2
MFCD00015650
Copper (II) ethoxide
C4H10CuO2
153.67
UN3181

Specifications

≈120°C (dec.)
25g
Air & Moisture Sensitive
Cu(OC2H5)2
MFCD00015650
Copper (II) ethoxide
2850-65-9
Green
C4H10CuO2
153.67
UN3181
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit