Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) fluoride, 98%, anhydrous

Catalog No. AC203371000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC203371000 100 g Plastic bottle
AC203370250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC203371000 Supplier Thermo Scientific Chemicals Supplier No. 203371000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7789-19-7 | CuF2 | 101.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-19-7
Molecular Formula CuF2
Molecular Weight (g/mol) 101.54
Synonym Cupric fluoride

Specifications

Melting Point 950.0°C
Color Blue-Gray to Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 97.5% min
Linear Formula CuF2
Merck Index 15,2628
Solubility Information Solubility in water: 47g/L (20°C). Other solubilities: hydrolyzes in hot water (forming oxyfluoride), soluble in acids and alcohol
Formula Weight 101.54
Percent Purity 98%
Loss on Drying 0.5% max.
Packaging Plastic bottle
Specific Gravity 4.23
Density 4.2300g/mL
Chemical Name or Material Copper(II) fluoride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
EINECSNumber 232-147-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.