Learn More
Strem, An Ascensus Company CAS# 544-19-4. 50g. Copper(II) formate hydrate. MFCD00150509
SDP
Catalog No. 5090114884
Encompass
CAS# 544-19-4. 50g. Copper(II) formate hydrate. MFCD00150509. Molecular Weight: 153.55. Molecular Formula: Cu(OOCH)2∙XH2O. Color/form: green to blue pwdr. Strem# 93-2925. http://www.strem.com/catalog/v/93-2925/
Catalog No. 50-901-14884 Supplier Strem, An Ascensus Company Supplier No. 93292550G
CAS# 544-19-4. 50g. Copper(II) formate hydrate. MFCD00150509. Molecular Weight: 153.55. Molecular Formula: Cu(OOCH)2∙XH2O. Color/form: green to blue pwdr. Strem# 93-2925. http://www.strem.com/catalog/v/93-2925/