Learn More
Medchemexpress LLC Copper(II) Gluconate | 527-09-3 | 98.0% | 453.84 | 1 ML
SDP
Catalog No. 5000367026
Encompass_Preferred
Copper(II) Gluconate is a non-toxic copper supplement aid and the copper salt of D-gluconic acid. It can be used as a precursor catalyst in the photo-induced polymerization of acrylates. This product is provided as a 10 mM solution in 1 mL of DMSO.
  • Can serve as a catalyst precursor for controlled polymerization of acrylate monomers.
  • Ligand exchange can be accelerated by ultraviolet irradiation or elevated temperature.
  • Suitable for in vitro research.
Catalog No. 50-003-67026 Supplier Medchemexpress LLC Supplier No. HYW01086010MM/1ML
Copper(II) Gluconate is a non-toxic copper supplement aid and the copper salt of D-gluconic acid. It can be used as a precursor catalyst in the photo-induced polymerization of acrylates. This product is provided as a 10 mM solution in 1 mL of DMSO.
  • Can serve as a catalyst precursor for controlled polymerization of acrylate monomers.
  • Ligand exchange can be accelerated by ultraviolet irradiation or elevated temperature.
  • Suitable for in vitro research.