Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(II) nitrate hemi(pentahydrate)

Catalog No. 46785550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-785-550G 50 g
46-785-5250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-785-550G Supplier Sigma Aldrich Supplier No. 46785550G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyzes oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material.

Specifications

CAS 19004-19-4
Molecular Formula CuN2O6 -+ 2.5H2O
Linear Formula Cu(NO3)2 -+ 2.5H2O
Quantity 50 g
MDL Number MFCD02253001
Synonym Cupric nitrate hemi(pentahydrate)
Molecular Weight (g/mol) 232.59
Percent Purity ≥99.99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.