Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(Ii) Nitrate Hemi(Pentahydrate), Solstice

Catalog No. 6000444 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-004-44 2.5 kg
60-004-45 500 g
60-004-43 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-004-44 Supplier Solstice Supplier No. 128372.5KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

≥98%,≥26% Cu basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19004-19-4
MDL Number MFCD02253001
Synonym dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate

Specifications

Physical Form Crystals
Quantity 2.5 kg
Packaging Plastic Bottle
Linear Formula Cu(NO3)2 · 2.5H2O
UN Number 3085
Molecular Weight (g/mol) 232.59
Formula Weight 232.59g/mol
Percent Purity ≥98%
Grade ACS
Chemical Name or Material Copper(Ii) Nitrate Hemi(Pentahydrate)
Hazard Statement P210-P260-P280-P305 + P351 + P338-P370 + P378-P371 + P380 + P375
Precautionary Statement H271-H290-H302-H314-H410
EINECSNumber 221-838-5
RTECSNumber QU7400000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.