Learn More
Strem, An Ascensus Company CAS# 10031-43-3. 25g. Copper(II) nitrate trihydrate (99.999%-Cu) PURATREM. MFCD00149671
SDP
Supplier:  Strem, An Ascensus Company 93294025G
Encompass
CAS# 10031-43-3. 25g. Copper(II) nitrate trihydrate (99.999%-Cu) PURATREM. MFCD00149671. Molecular Weight: 187.56 (241.60). Molecular Formula: Cu(NO3)2∙3H2O. Color/form: blue xtl. Strem# 93-2940. http://www.strem.com/catalog/v/93-2940/
Catalog No. 50-901-14895