Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (II) Nitrate, Trihydrate
SCI_ED

Catalog No. S25282A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
Reagent Grade
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25282A 500 g Reagent Grade
S25282 100 g Reagent Grade
S25282B 500 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25282A
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10031-43-3
Molecular Formula CuH6N2O9
Molecular Weight (g/mol) 241.599
InChI Key SXTLQDJHRPXDSB-UHFFFAOYSA-N
Synonym copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
PubChem CID 9837674
IUPAC Name copper;dinitrate;trihydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]

Specifications

Color Blue
Quantity 500 g
Linear Formula Cu(NO3)2·3H2O
Formula Weight 241.6
Packaging Reagent Grade
Density 2.05g/mL
Chemical Name or Material Copper (II) Nitrate, Trihydrate
Safety handling Check Alert storage code Red
ShelfLife 1 year
DOTInformation DOT Class 5.1, II, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.