Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Copper(II) nitrate trihydrate, 99%, for analysis
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                100 g
                            
                        
                            
                                500 g
                            
                        
                            
                                2.5 kg
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Plastic bottle
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
            Specifications
Specifications
| Melting Point | 114.0°C | 
| Color | Brown to Cream | 
| Physical Form | Fine Powder | 
| Quantity | 500 g | 
| Assay Percent Range | 98.5 to 104% (Iodometry) | 
| Molecular Formula | CuN2O6·3H2O | 
| Linear Formula | Cu(NO3)2·3H2O | 
| Fieser | 17,366 | 
| Merck Index | 15, 2633 | 
| Solubility Information | Solubility in water: 267 g/100 mL. Other solubilities: freely soluble in alcohol, practically insoluble in ethyl acetate | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Danger | 
| Hazard Statement | GHS H Statement May intensify fire; oxidizer. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. | 
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove conta | 
| RTECSNumber | GL7875000 | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                