Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) nitrate trihydrate, 99%, for analysis

Catalog No. p-46599
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207681000 100 g Plastic bottle
AC207685000 500 g Plastic bottle
AC207680025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC207681000 Supplier Thermo Scientific Chemicals Supplier No. 207681000
Only null left
Add to Cart

CAS: 10031-43-3 | CuN2O6·3H2O | 241.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-43-3
MDL Number MFCD00149671
InChI Key SXTLQDJHRPXDSB-UHFFFAOYSA-N
Synonym copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
PubChem CID 9837674
IUPAC Name copper;dinitrate;trihydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]

Specifications

Melting Point 114.0°C
Color Brown to Cream
Physical Form Fine Powder
Quantity 100 g
Assay Percent Range 98.5 to 104% (Iodometry)
Molecular Formula CuN2O6·3H2O
Linear Formula Cu(NO3)2·3H2O
Fieser 17,366
Merck Index 15, 2633
Solubility Information Solubility in water: 267 g/100 mL. Other solubilities: freely soluble in alcohol, practically insoluble in ethyl acetate
Molecular Weight (g/mol) 241.6
Formula Weight 241.6
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Copper(II) nitrate trihydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
RTECSNumber GL7875000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.