Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (II) Oxide
SCI_ED

Catalog No. S25284 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Reagent Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25284 25 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25284
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
IUPAC Name copper(2+) oxidandiide
SMILES [O--].[Cu++]

Specifications

Quantity 25 g
Linear Formula CuO
Formula Weight 79.54
Grade Reagent
Packaging Reagent Grade
Density 6.315g/mL
Chemical Name or Material Copper (II) Oxide
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.