Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) oxide, Solstice

Catalog No. 6000450 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-004-50 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-004-50 Supplier Solstice Supplier No. 61200500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a.,≥99.0% (RT), powder

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
IUPAC Name oxocopper
SMILES [O--].[Cu++]

Specifications

Physical Form Powder
Quantity 500 g
UN Number UN3077
Formula Weight 79.55g/mol
Percent Purity ≥99% (RT)
Packaging Plastic Bottle
Chemical Name or Material Copper(II) oxide
Hazard Statement P273
RTECSNumber GL7900000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.