Learn More
Sigma Aldrich Fine Chemicals Biosciences Copper(II) oxide on alumina 14-30 mesh, extent of labeling: 13 wt. % loading | Purity: 9.0-11.0% Cu basis (EDTA titration) | Mol Wt: 79.55 | 1317-38-0 | MFCD00010979 | 250G
Catalog No. 501881061
Encompass
Copper(II) oxide on alumina 14-30 mesh, extent of labeling: 13 wt. % loading | Purity: 9.0-11.0% Cu basis (EDTA titration) | Mol Wt: 79.55 | 1317-38-0 | MFCD00010979 | 250G
Catalog No. 50-188-1061 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 417971250G
Copper(II) oxide on alumina 14-30 mesh, extent of labeling: 13 wt. % loading | Purity: 9.0-11.0% Cu basis (EDTA titration) | Mol Wt: 79.55 | 1317-38-0 | MFCD00010979 | 250G