Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) Oxide Wire, For Microanalysis, MilliporeSigma™

Catalog No. M1027625000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1027625000 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1027625000 Supplier MilliporeSigma Supplier No. 1.02762.5000

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
IUPAC Name copper(2+) oxidandiide
SMILES [O--].[Cu++]

Specifications

Appearance Dark gray wire
Quantity 5 kg
UN Number UN 3077
Formula Weight 79.55
Odor Odorless
Packaging Plastic bottle
Form Wire
Chemical Name or Material Copper(II) Oxide
Hazard Category H302: Harmful if swallowed.
H410: Very toxic to aquatic life with long lasting effects.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.