Learn More
Medchemexpress LLC Copper(II) perchlorate hexahydrate | 10294-46-9 | 149666 | 98.0% | 370.52 g/mol | Cu(ClO4)2·6H2O | 250 G
SDP
Catalog No. 5000209597
Encompass_Preferred
Copper(II) perchlorate hexahydrate (Cu(ClO4)2·6H2O) is a water-soluble inorganic copper salt supplied as the hexahydrate. It is used as a soluble Cu(II) source in synthetic chemistry, coordination chemistry, and in catalytic or oxidative reactions where a perchlorate counterion is acceptable.
  • Provides a soluble source of copper(II) for synthesis and catalysis.
  • Hexahydrate form gives a defined hydration state for stoichiometric calculations.
  • Suitable for coordination chemistry and reagent preparation.
  • Available in laboratory-scale packaging for research use.
  • Perchlorate salts are oxidizing; handle with appropriate safety precautions.
Catalog No. 50-002-09597 Supplier Medchemexpress LLC Supplier No. 250GHYW075791
Copper(II) perchlorate hexahydrate (Cu(ClO4)2·6H2O) is a water-soluble inorganic copper salt supplied as the hexahydrate. It is used as a soluble Cu(II) source in synthetic chemistry, coordination chemistry, and in catalytic or oxidative reactions where a perchlorate counterion is acceptable.
  • Provides a soluble source of copper(II) for synthesis and catalysis.
  • Hexahydrate form gives a defined hydration state for stoichiometric calculations.
  • Suitable for coordination chemistry and reagent preparation.
  • Available in laboratory-scale packaging for research use.
  • Perchlorate salts are oxidizing; handle with appropriate safety precautions.