Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(II) phthalocyanine

Catalog No. 502908322
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-8322 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-290-8322 Supplier Sigma Aldrich Supplier No. 5466745G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Copper(II) phthalocynaine (CuPc) is a metal phthalocyanine dye that acts as a p-type semiconductor. It has a charge mobility of 10°C to 4°cm2/Vs. It forms a chemically stable thin film that exhibits photoconductivity and catalytic activity.CuPc is a semiconducting material that can be used in a variety of optoelectronic devices such as light emitting diodes (LEDs), field effect transistors (FETs) and solar cells/organic photovoltaics (OPVs).
TRUSTED_SUSTAINABILITY

Specifications

CAS 147-14-8
Molecular Formula C32H16CuN8
Linear Formula C32H16CuN8
Quantity 5 g
MDL Number MFCD00010719
Synonym CuPc; Phthalocyanine blue; Pigment Blue 15
Molecular Weight (g/mol) 576.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.